Learn More
McKesson General Medical Sterilization Bowie-Dick Test Pack Steam, 30/cs
Supplier: McKesson General Medical 724430
Easy-to-read test sheet clearly identifies failures and can be filed for record keeping. Test pack will reproducibly detect residual air in the sterilizer chamber due to air leaks, air removal failures and inadequate vacuum by testing the air removal efficiency of 132-134˚C (270-273˚F) dynamic air-removal steam sterilizers. Indicator sheet will turn a uniform dark brown/black color except when air removal failures, such as air leaks, occur; can be filed for record keeping.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.