Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DR Instruments Botany Dissecting Kit
SDPSCI_ED

Economical dissecting kit for botany students, teachers and field botanists.

Supplier:  DR Instruments 10BPC

Catalog No. S06838

Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative.


Includes:
  • Dissecting Iris scissors, 4.5 in.
  • Two dissecting forceps, 4.5 in. with guide pin (one curved fine point, one medium point)
  • Two plastic teasing needles (one angular and one straight)
  • Dissecting scalpel handle No. 3
  • No.10 blade (10 pcs.) or No. 15 blade
  • Ruler, 6 in.
  • Glass dropper
  • Color plastic case

Description

Description

  • For both field work and lab use
  • Superior quality dissection tools available in washable, sanitizable plastic case
  • Contains all the necessary tools required for plant study, dissection and analysis
  • All tools are made with stainless steel, unless otherwise noted

Specifications

Specifications

1 Ea.
Basic Botany Dissection Kit
Economical dissecting kit for botany students, teachers and field botanists
Stainless Steel
4.5 in. dissecting iris scissors, 4.5 in. two dissecting forceps, with guide pin, two teasing needles (one angular, one straight) No. 3 dissecting scalpel handle, No. 10 blade (10 pcs.) or No. 15 blade, 6 in. ruler, glass dropper, color plastic case
Vinyl
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit