Learn More
Fine Science Tools Delicate Bone Scraper, Stainless Steel, 16.5 cm
Supplier: Fine Science Tools 1007516
Tip Shape: Angled/ Cutting Edge: 0.4mm/ Alloy / Material: Stainless Steel/ Length: 16.5cm. - The edges of this instrument are medium sharp so that they can scrape tissue from delicate bone without cutting into the bone itself.