Learn More
Fine Science Tools Bone Cutting Spring Scissors, Stainless Steel, 13 cm
Supplier: Fine Science Tools 1614413
Tip Shape: Straight/ Tips: Blunt/ Cutting Edge: 12mm/ Tip Diameter: 0.4mm/ Alloy / Material: Stainless Steel/ Length: 13cm/ Serrated: No