Learn More
Boekel Scientific™ Open Source Bio™ (OSB) Transmitter with pH, DO and Conductivity Probes
A flexible pH, DO and Conductivity measuring device.
Supplier: Boekel Scientific™ 555004/EMD
Includes: Open Source Transmitter, pH probe, Conductivity probe, DO probe and calibration reagents.
Description
Open Source Bio or OSB probes and transmitters work just like conventional probes and transmitters but allow for additional functionality because the equipment is built around an Arduino™ compatible board. The transmitter is capable of single parameter measurement of pH, DO and Conductivity. The transmitter includes high precision temperature probe for temperature compensation.
- Built around UNO, an open source circuit board that allows easy user programming and experiment control
- High precision temperature probe for temperature compensation
- Repairable and industrial design supports years of experiments
- External I/O connector for connection to pumps or alarms
- User friendly probe calibration menus
- USB drive data logging
- Large color screen
Specifications:
- pH Range: 0.001 to 14.000
- Conductivity Range: 0.07 us/cm to 500,000 us/cm
- DO Range: 0.01 to 39.99mg/L
- Accuracy: ±0.02 (pH), ±2us/cm (conductivity), ±0.02 (DO)
- Calibration: 2-/3-Point
Warranty and Services
Two years.
Specifications
pH: ±0.02±; Conductivity: ±2us/cm; DO: ±0.02 | |
±0.02 | |
2 or 3 | |
Open Source Transmitter, pH probe, Conductivity probe, DO probe and calibration reagents | |
pH: 0.001 to 14.000; Conductivity: 0.07 us/cm to 500,000 us/cm; DO: 0.01 to 39.99mg/L |
±2us/cm | |
±0.02 | |
Large Screen | |
OSB Transmitter with three probes (pH, conductivity, DO) | |
2 Years |