Learn More
Boekel Scientific™ Open Source Bio™ (OSB) Transmitter with Conductivity Probe
A flexible device for measuring conductivity.
Supplier: Boekel Scientific™ 555002/EMD
Includes: Open Source Transmitter, conductivity probe, calibration reagents and temperature probe.
Description
Open Source Bio or OSB probes and transmitters work just like conventional probes and transmitters but allow for additional functionality because the equipment is built around an Arduino™ compatible board. The transmitter is capable of single parameter measurement of pH, DO and Conductivity. The transmitter includes high precision temperature probe for temperature compensation.
- Built around UNO, an open source circuit board that allows easy user programming and experiment control
- High precision temperature probe for temperature compensation
- Repairable and industrial design supports years of experiments
- External I/O connector for connection to pumps or alarms
- User friendly probe calibration menus
- USB drive data logging
- Large color screen
Specifications:
- Range: 0.07us/cm to 500,000us/cm
- Accuracy: ±2us/cm
- Calibration: 2-Point
- Also works with DO and pH probes
Warranty and Services
Two years.
Specifications
2-Point | |
±2us/cm | |
Large Screen | |
Open Source Transmitter, conductivity probe, calibration reagents and temperature probe | |
2 Years |
2 | |
0.07 us/cm to 500,000 us/cm | |
±2us/cm | |
OSB Transmitter with EC supplies and Temperature probe |