Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Block, 24 x 0.2 mL, EMD
For Use With: Thermo Scientific™ Compact Dry Bath/Block Heater
Supplier: Thermo Scientific™ 88871101/EMD
Description
Increase your application capabilities with a variety of accessories for the Thermo Scientific™ Compact Dry Baths/Block Heaters.
Warranty: 2 yearsSpecifications
24 x 0.2mL block | |
24 | |
36 x 54 x 15 mm (1.4 x 2.1 x 0.6 in.) |
Thermo Scientific™ Compact Dry Bath/Block Heater | |
Block |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction