Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Agilent BioTek™ BioSpa 8 Automated Incubator PROMO
The BioTek™ BioSpa live cell analysis system offers unique benefits for a wide variety of live cell imaging applications in up to 8 microplates or other labware.
Supplier: Agilent BioTek™ BIOSPAGSN2358
This item is not returnable.
View return policy
Description
With 5 imaging modes, the BioTek BioSpa live cell analysis system enables a wide range of microscopy applications.
- Capture detailed cellular images, get cell-level analyses for migration and wound healing assays, 3D spheroid formation, and many other kinetic applications
- Built-in scheduling, environmental monitoring, and available liquid handling for walk away automation
- Multiple users can run simultaneous processes for maximum efficiency in the lab
Note: For Research Use Only. Not for use in diagnostic procedures.
Warranty and Services
1 Year
Specifications
Ambient to 45°C | |
20.1 in. | |
18.9 in. | |
80 to 95% | |
27.2 in. | |
Automated Microplate Incubator |
CE and TUV Marked, RoHS Compliant | |
51 cm | |
48 cm | |
CO2/O2 Control and Monitoring, Water Pan and Humidity Monitoring, BioSpa Software, One Alignment Kit | |
69 cm | |
8 Plates |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction