Learn More
Biomechanical Kits
Offer a complete lab-tested system for teaching and learning the musculoskeletal system
Supplier: Denoyer Geppert Company 065012
Includes:
All Kits Include:
- Support base and rod
- Support clamps
- Force cord assembly (cable, handle, connectors, fine adjustment, force gauge)
- instructor's curriculum guide
- Short curriculam for introductoray A&P courses
- Student Laboratory guide
Biomechanical Arm Kit Only
- Biomechanical arm model
- Brass weights (1-50g, 1-100g)
Biomechanical Leg Kit Only
- Biomechanical leg model
- Brass weights (2-200g)
Complete Biomechanics Module
- Contains everything listed above
Description
By mastering a limited set of basic concepts, students can predict and explain body movement using the same approaches employed by kinesiologists, exercise physiologists, physical therapists, and orthopedic nurses. Working ideally in teams of four, students practice observation, measurement, data gathering and analysis, prediction, plus calculations and problem solving.
Order Info
Some kit components also sold individually.
Specifications
Biomechanical Leg Kit | |
For teaching and learning the musculoskeletal system | |
Biomechanical leg model, support base, rod and clamps, force cord assembley (cable, handle connectors, fine adjustment force gauge), brass weights (2-200 g), instructor's curriculum guide and four student laboratory guides | |
Support Base and Rod |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.