Learn More
Quantimetrix Bilirubin Control Pediatric, Level 1
Supplier: Quantimetrix 133131
• Neonatal bilirubin levels up to 20 mg/dL• Liquid, human protein matrix• 2-year refrigerated shelf life from date of manufacture• 21-day open vial stability when stored at 2ºC–8ºCMonitor the performance of neonatal bilirubin assay methods including Reichert Unistat and Advanced Instruments BR2 Bilirubin Stat-Analzer Model bilirubinometers and all automated bilirubin methods for popular instruments including Abbott Architect, Beckman Coulter AU Series, Roche Cobas, Siemens Advia and Siemens Dimension.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.