Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3B Scientific™ Biface Replica
SCI_ED

3B Scientific™ Biface Replica is an accurate reproduction of a biface made from quartzite

Supplier:  3B Scientific™ U75030

Catalog No. S20737


Add to Cart

Description

Description

  • Original finding likely used by an ancestor of the Neanderthals as a multi-purpose tool
  • Material: Liquid wood
  • Age: 0,3 - 0,4 Mya
  • Place found: Neandertal
Specifications

Specifications

Model
19 cm
Fossil
English
10 cm
1
Gray
Earth Science
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit