Learn More
Berlese Apparatus
Simple and effective model for collecting nematodes and microarthropods from soil samples.
Supplier: Neo/Sci Corporation 301040
Includes: Comprehensive teacher and student guides
Description
To Use:
- Place soil sample in top portion, then cover with lamp
- Light's heat desiccates the soil
- Organisms migrate down toward the attached collection jar for study or preservation
Specifications
Berlese Apparatus | |
A simple, effective and educational means of gathering nematodes and microarthropods from soil samples | |
27.9 cm |
11 in. | |
Teacher's guide |