Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Benzoic Acid Copper(II) Salt Hydrate 96%, Thermo Scientific™
Click to view available options
100g; Glass bottle
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
CAS | 66019-07-6 |
CAS Min % | 95.0 |
CAS Max % | 100.0 |
Assay Percent Range | 96% |
Molecular Formula | C14H10CuO4·xH2O |
Linear Formula | (C6H5CO2)2Cu·xH2O |
MDL Number | MFCD03066124 |
Formula Weight | 305.77 |
Percent Purity | ≥95% |
Quantity | 100g |
Show More |
Safety and Handling
Hazard Statement | GHS H Statement: Harmful if swallowed. |
Precautionary Statement | GHS P Statement: IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
EINECSNumber | 208-552-6 |
TSCA | TSCA |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction