Learn More
Pendotech Pendotech Benchtop Photometer With 280Nm Light Source
Supplier: Pendotech SPECL1SU280
Benchtop Photometer w/280nm light source, 4-20mA output, (2) 1 meter fiber optic cables with tethers, (2) optical couplers with tethers to connect to flow cell, 24VDC power supply