Learn More
Bel-Art™ SP Scienceware™ Vacuum Pressure Indicator
Indicates if desiccator is under vacuum.
Supplier: Bel-Art™ H420420000
Description
- Thermoplastic elastomer collapses with negative vacuum pressure in desiccator and expands when vacuum is absent
- Fits onto tapered end of stopcock
Specifications
Thermoplastic elastomer bellows collapses when under negative pressure (vacuum), expands when under ambient atmospheric pressure, indicates the desiccator vacuum | |
Fits on to tapered end of stopcock | |
Vacuum-Pressure Indicator |