Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bel-Art™ SP Scienceware™ Compartment Trays
Protect pipets, thermometers and any other long, slim items you want to protect from breakage.
Supplier: Bel-Art™ H186480000/EMD
Description
- Designed to hold pipets up to 16.1 in. (41cm)
- Two compartments 4 × 16.1 in. (10 × 41cm) and one compartment 12 × 41cm
- All compartments 2.5 in.H (6.4cm)
Specifications
Compartment Tray | |
6.4 cm | |
Polystyrene |
2.5 in. | |
White |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction