Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HemoCue America Beckman Coulter™ ICON™ HP Test Kit
Easy, fast and accurate results
Supplier: HemoCue America 395160A
Includes: 30 Test strips, 35 capillary tubes, 1 dropper bottle of developer solution containing 0.1% Sodium Azide, product instructions and procedure card
Description
- CLIA Complexity: Waived (whole blood); moderate (serum/plasma)
- Rapid anti-H. pylori antibody test uses indirect solid-phase immunoassay technology
- Detects anti-Helicobater pylori IgG antibody in human whole blood, serum or plasma specimens
- One-step cassette device test aids in diagnosis of H. pylori infection in adults with gastrointestinal symptoms
- Results in 10 minutes or less
- Room temperature storage
Specifications
Waived, Non-waived | |
H. pylori IgG Test Kit | |
30 Tests |
H. pylori Antibodies | |
30 Test strips, 35 capillary tubes, 1 dropper bottle of developer solution contains 0.2% Sodium Azide, reagents, product instructions, procedure card |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction