Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HemoCue America Beckman Coulter™ ICON™ DS Strep A Test
Easy-to-use dipstick format
Supplier: HemoCue America 395098A
Description
- CLIA Complexity: Waived
- Qualitative assay for rapid detection of Strep A antigen from throat swabs
- Results in five minutes
- Sensitivity: 97%; specificity: 95%
Specifications
Waived | |
Throat Swab | |
25 Test devices, 25 extraction tubes, 25 sterile swabs, 1 extraction reagent A and B, 1 positive and negative control, 1 product instructions and procedure card | |
5 min. |
Streptococcal Antigen (Group A) | |
Strep A Test Kit | |
25 Pack |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction