Learn More
BD Golgi Sampler Kit
It is involved in the sorting of nascent proteins into carrier vesicles for transport to appropriate destinations
Supplier:  BD 611434
Catalog No. BDB611434
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
  • Golgi apparatus is a collection of flattened membranes, or cisternae, and vesicles
  • Protein maturation and post translational modification occur after their biosynthesis at the ER and transport through the Golgi apparatus
  • This involves the transport of protein containing vesicles through a vectorial process of vesicle budding and fusion from the cis-compartment (CGN) to the medial-compartment (MGN) and the trans-compartment (TGN) of the Golgi apparatus. Various proteins resident to specific Golgi compartments are necessary for proper function
  • TGN38 is a type I integral membrane protein primarily localized to the TGN
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Store undiluted at -20°C
Western blot
10 μg
Golgi Sampler Kit
Recommendations
Recommendations