Learn More
BD Staphyloslide™ Latex Test Kit
Test results within 20 seconds
Supplier: BD 240952
Includes: Required reagents, disposable test cards, and stirrers.
Description
- CLIA Complexity: High
- Rapid, latex slide agglutination test for detecting a cell-wall polypeptide clumping factor (CF) produced by most S. aureus strains
- Test consists of sheep red blood cells sensitized with fibrinogen
- Organisms possessing clumping factor cause clumping of erythrocytes
Specifications
Cell wall polypeptide clumping factor, Protein A (S. aureus strains) | |
StaphyloSlide™ Latex Test Kit | |
Required reagents, disposable test cards and stirrers | |
100 Tests |