Learn More
BD Phospholipid-binding Protein, APC Annexin V
Has high affinity for PS, and binds to cells with exposed PS
Supplier:  BD 561012
 View more versions of this product
Catalog No. BDB561012
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
  • Conjugated to APC
  • Staining precedes the loss of membrane integrity which accompanies the latest stages of cell death resulting from either apoptotic or necrotic processes
Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
25 Tests
Aqueous Buffered solution containing BSA and ≤0.09% Sodium Azide
Flow Cytometry
5 μL
4°C
APC (Allophycocyanin)
Product Suggestions
Product Suggestions