Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD DRAQ7™
Used to stain nucleic acids in fixed cells. BD Pharmingen™ DRAQ7™ is also used for cell cycle analysis by DNA content, nuclear visualization, or discrimination of nucleated cells from debris or neucleated cells.
Supplier: BD 564904
Description
- Far-red fluorescent DNA dye
- Cell impermeable
- Used to stain nucleic acids in fixed cells
- Has an excitation wavelength maxima of 599/644nm
Specifications
Far-red | |
Store undiluted at 4°C | |
DNA dye | |
|
0.3mM | |
599/644nm | |
|
|
1mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction