Learn More
BD FACSArray™ Daily QC Particles
Consist of three discrete fluorescent or non-fluorescent particles for daily instrument quality control to confirm consistent instrument performance
Manufacturer: BD 558542
Description
The SPHERO™ Daily QC Particles for BD FACSArray™ are useful for daily testing of BD FACSArray bioanalyzer to confirm consistent instrument performance and have been tested on the BD FACSArray for this purpose. The %CV of rainbow calibration particles (Peak 8) should always be ≤7% in all fluorescent parameters.
- Each particle contains mixture of fluorophores
- Excitation wavelength range: 365 - 650nm
- Storage buffer: Aqueous solution containing 0.01%NP40 and ≤0.02% sodium azide
- Rainbow calibration particle: Peak 8 (Bright)
- Rainbow calibration particle: Peak 4 (Dim)
- Blank calibration particle: Peak 1 (Negative)
Flow cytometry
Specifications
Flow Cytometry | |
Fluorescent or Non-fluorescent | |
3 Vials |
Safety and Handling
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Research use only.