Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: UVM Modified Listeria Enrichment Broth
Selective enrichment for culturing L. monocytogenes
Supplier: BD 222330
Description
Specifications
AOAC, CCAM, COMPF, USDA | |
Dehydrated | |
Microbiology Media | |
For rapidly isolating Listeria monocytogenes | |
500 g |
Refrigerate | |
Listeria Enrichment Broth | |
Pancreatic digest of Casein 5g/L, Proteose Peptone No. 3 5g/L, Beef extract 5g/L, Yeast extract 5g/L, Sodium Chloride 20g/L, Disodium Phosphate 9.6g/L, Monopotassium Phosphate 1.35g/L, Esculin 1g/L, Nalidixic Acid 0.02g/L, Acriflavine HCl 12mg/L | |
Bottle |
Safety and Handling
ShelfLife : 1095 Days
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction