Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Tellurite Solution

Sterile 1% solution of potassium tellurite, a differential agent

Supplier:  BD 215294

Catalog No. B215294


Edge
Add to Cart
This item is not returnable. View return policy

Description

Description

  • Prepared, dehydrated culture media
  • Used as an additive
  • Coagulase-positive staphylococci reduce tellurite and produce black colonies
  • Available in 5mL tubes

Specifications

Specifications

2° to 8°C
Media Supplement
Sterile
5 mL
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit