Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Tigecycline

High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents

$238.50

Specifications

Antimicrobial Agents Tigecycline
Concentration 15μg
Content And Storage 2°C to 8°C
Packaging Type Disc
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
B232087
View Documents
BD
232087
1/Pk.
Each for $49.35
N/A
Due to product restrictions, please to purchase or view availability for this product.
14-959-901
View Documents
BD
232208
10/Pk.
Pack of 10 for $238.50
Edge
Add to Cart
 
Description

Description

  • Clearly marked on both sides with letters and numbers designating agent and concentration

Specifications

Specifications

Tigecycline
15μg
2°C to 8°C
Disc
Safety and Handling

Safety and Handling

ShelfLife : 365 Days

SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit