Learn More
Cole-Parmer Newport LIP275-1222-01 Balance Isolation Platform, 12" W x 22" D
Supplier:  Cole-Parmer 1150003
Encompass
Lightweight and easy to set up, the platform isolates balances and other sensitive instruments and equipment from vibrations and mechanical shock—ideal for balances, petri dishes, and shakers. Sturdy, gray PVC platform features a non-stick neoprene surface supported by three or four vibration isolation bearings. These advanced isolation bearings protect balances and other sensitive equipment from ambient vibrations and mechanical shocks.
Catalog No. NC1569844