Learn More
FUJIFILM IRVINE SCIENTIFIC INC BalanCD CHO Growth A Medium - Powder, 10 L
Supplier: FUJIFILM IRVINE SCIENTIFIC INC 9412010L
BalanCD CHO Growth A is a chemically-defined, animal component-free growth medium designed to increase process yields of antibodies and recombinant proteins in Chinese Hamster Ovary (CHO) cells. This medium is designed to be used in conjunction with BalanCD CHO Feed 4 for optimal performance in fed-batch cultures.