Learn More
FUJIFILM IRVINE SCIENTIFIC INC BalanCD CHO Feed 2 - Liquid, 1 L
Supplier: FUJIFILM IRVINE SCIENTIFIC INC 911291L
BalanCD CHO Feed 2 is a chemically-defined, animal component-free feed medium designed to increase process yields of antibodies and recombinant proteins in Chinese Hamster Ovary (CHO) cells in fed-batch mode. BalanCD CHO Feed 2 was designed to be used with a wide range of growth media for optimal performance in fed-batch cultures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.