Learn More
Microtech Scientific BACTI BOTTLE 120ML PP 200/CS
Supplier:  Microtech Scientific MS5R120PP
Encompass_Preferred
Water Sample Bottles, 120mL Polypropylene, with Sodium Thiosulfate and STB (Breakaway) Tamper Caps, Sterile, 100/Box, 200/Case
Catalog No. NC9712005