Learn More
Waters Corp BACKPRESSURE REGULATOR, 250PSI
Supplier: Waters Corp 700002676
Backpressure Regulator, 250psi;ACQUITY UPLC PDA Detector,ACQUITY UPLC eLambda PDA Detector,ACQUITY UPLC Fluorescence Detector,ACQUITY UPLC TUV Detector