Learn More
Selleck Chemical LLC Azacitidine (5-Azacytidine) 10mM/1mL 320-67-2 5-AzaC,Ladakamycin, AZA,5-Aza
Supplier: Selleck Chemical LLC S178210MM/1ML
Azacitidine (5-Azacytidine, 5-AzaC, Ladakamycin, AZA, 5-Aza) is a nucleoside analogue of cytidine that specifically inhibits DNA methylation by trapping DNA methyltransferases. Azacitidine induces mitochondrial apoptosis and autophagy. *For Research & Development use only. Product is not intended for drug, household, or human consumption.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.