Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Axygen™ Multimek Robotic Tips: 200μL
Fits Multimek «Type II« or CCS-style head
$974.00
Specifications
Volume (Metric) | 200 μL |
---|---|
Filtered | Non-Filtered |
Color | Clear |
Rack Color | Blue |
For Use With (Application) | Unique diamond polished molding technology use no mold-releasing agents, minimizes wastage of precious samples and reagents while handling, nucleic acid, nuclease and endotoxin-safe for complete protection against contamination |
Catalog Number | Mfr. No. | Sterility | Tip Style | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Sterility | Tip Style | Price | Quantity & Availability | ||||
14-222-936
|
Axygen™
SCT200SSBRS |
Non-sterile | Automation |
Case of 4000 for $974.00
|
|
||||
14-222-937
|
Axygen™
SCT200SSGRS |
Sterile | Automation |
Case of 4000 for $974.00
|
|
||||
Description
Robotic tip's unique diamond-polished molding technology uses no mold-releasing agents. Minimizes wastage of precious samples and reagents while handling. Nucleic acid, nuclease and endotoxin-safe for complete protection against contamination.
- Tip volume: 200μL
- Tip color: Clear
- Rack color: Red and blue
Order Info
Racked: 96 tips/rack, 10 racks/pack, 5 packs/case
Specifications
200 μL | |
Clear | |
Unique diamond polished molding technology use no mold-releasing agents, minimizes wastage of precious samples and reagents while handling, nucleic acid, nuclease and endotoxin-safe for complete protection against contamination | |
Automation |
Non-Filtered | |
Blue | |
Rack | |
Beckman Coulter Multimek |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title