Learn More
IBA LifeSciences Avidin; 50 mg
Supplier: IBA LifeSciences 20204050
Avidin is a tetrameric biotin binding protein (KD ~1 x 10-14) extracted from egg white. It prevents biotin and biotinylated proteins from binding to Strep-Tactin or Strep-TactinXT, whereas binding of the Twin-Strep-tag and Strep-tagII is not influenced. Biotin and/or biotinylated proteins can potentially reduce the binding capacity of Strep-Tactin as well as Strep-TactinXT and are present in cell lysates and cell culture media, especially from mammalian or insect cells. Thus, if Twin-Strep-tag and Strep-tagII fusion proteins from biotin-containing extracts or media are intended to be purified via Strep-Tactin or Strep-TactinXT affinity chromatography, biotin has to be masked by addition of avidin prior to application onto the resin.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.