Learn More
Rockland Immunochemicals AVIDIN - A003-01
Supplier: Rockland Immunochemicals A00301
Avidin is biotin-binding protein found in the oviducts of egg-laying animals (birds, reptiles, and frogs) that gets deposited into the whites of their eggs. Avidin is a tetramer and can bind up to four biotin molecules (Vitamin B7) with one of the greatest known non-covalent interactions. Avidity for biotin is destroyed with heat.