Learn More
Beckman Coulter Beckman Coulter Avanti J-15 Cell Culture Pkg
Supplier: Beckman Coulter C19417
Cell Culture Package: Avanti J-15, IVD, Ventilated Benchtop Centrifuge, 100-120 V, 50/60 Hz, JS-4.750 (4 x 750 mL) swinging bucket rotor, 15 mL conical tube adapters (qty 4) and 50 mL conical tube adapters (qty 4). (Bucket covers sold separately)
This item is not returnable.
View return policy