Learn More
Beckman Coulter Beckman Coulter Avanti J-15 Blood Pkg
Supplier: Beckman Coulter C19423
Blood Sample Package: Avanti J-15, IVD, Ventilated Benchtop Centrifuge, 100-120 V, 50/60 Hz, JS-4.750 (4 x 750 mL) swinging bucket rotor, 13 mm diameter tube adapters (qty 4) and 16 mm diameter tube adapters (qty 4). (Bucket covers sold separately)
This item is not returnable.
View return policy