Learn More
Cytiva AutoScreen 96-Well Filter Plates
For purification of sequencing reactions prior to analysis on MegaBACE 1000 and can be used for other size exclusion applications
Supplier:  Cytiva 25900598
Catalog No. 09-928-186
Add to Cart
Description
Description
AutoScreen 96-Well Plate consists of a 96-well filter plate containing DNA Grade Sephadex G-50 for purification of sequencing reactions prior to analysis on MegaBACE 1000 and can be used for other size exclusion applications.
Specifications
Specifications
Standard
8 x 12
96
Clean up prior to analysis on ABI sequencers