Learn More
Santa Cruz Biotechnology AUTORADIOGRAPHY FILM BLUE 8X10
SDP
Supplier:  Santa Cruz Biotechnology SC201697
Encompass
UltraCruz Autoradiography Film, Blue, 8 in. x10 in.. Autoradiography (X-ray) film is tinted with the classic blue color, suitable for all common uses including radioisotopes and chemiluminescent substrates. Autoradiography (X-ray) film creates accurate physical records of experimental results through enhanced chemiluminescence (ECL) with Western Blotting Luminol Reagent : sc-2048. Autoradiography film is ideal for creating precise images, scanning into a digital format for presentation, and publication. Applications: Western blotting, Northern & Southern blotting 20.3 cm x 25.4 cm (8 in. x 10 in.) 100 sheets per case
Catalog No. NC0357566