Learn More
Fine Science Tools AutoClip System
Supplier: Fine Science Tools 1202209
AutoClip System, Autoclave Safe: Yes, Sterile: No, Length: 0.9cm, Item: Clips, The AutoClip System is a safe, secure alternative method of wound closure. Faster and easier than suture, this system is perfect when staples may be too thin to provide adequate closure. Components sold separately or as a kit.