Learn More
Cole-Parmer Autoclave Deodorizers, Apple; 100/Bottle
Supplier: Cole-Parmer 2440674
These sterilizer deodorizers chemically absorb odor-causing molecules, removing unwanted odors from your facility rather than masking them. Simply place one capsule in your autoclave or incubator with each load. The heat and moisture dissolves the capsule and releases a clean, fresh scent.