Learn More
Lab-Aids™ Atom Modeling: Structure, Valence, & Bonding
SCI_ED
During the three investigations in this Lab-Aids Atom Modeling: Structure, Valence, & Bonding kit, students explore the structure of atoms and how this influences the properties of elements. Although atoms cannot be chemically split, there are smaller subatomic particles within each atom.
Manufacturer:  Lab-Aids™ 145N
Catalog No. S98162
Add to cart
Description
Description
  • Using an innovative model, students discover the basic arrangement of protons, neutrons, and electrons in atoms of a given element.
  • They then use the model to provide insight into why each element has certain properties and the patterns inherent in the Periodic Table.
  • Protons and neutrons are found in the nucleus
  • Electrons are found in an electron cloud surrounding the nucleus.
  • The atomic number is equal to the number of protons (or electrons) in an atom
  • The atomic mass equals the sum of the protons and neutrons
  • Atoms of different elements are distinguished by the number of protons in the nucleus
  • Isotopes of an element have different number of neutrons in the nucleus of their atom
  • The number and arrangement of subatomic particles influence the patterns of the Periodic Table of Elements
  • Valence electrons play a key role in bonding
Specifications
Specifications
24 Atom Model Boards, 16 Periodic Tables cards, 8 sets of subatomic particles (40 red, 40 blue, and 40 white spheres), 1 Teacher's Guide, 32 Student Guides
Science Kit