Learn More
ASTM™ D6584 Glycerin Solution, Certified Reference Material, 500 μg/mL in Pyridine, MilliporeSigma™ Supelco™
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Manufacturer:  MilliporeSigma Supelco CRM44892
Catalog No. 11-101-7905
Add to cart
Description
Description
Specifications
Specifications
56-81-5
MFCD00004722
Glycerol solution; Biodiesel impurity; 1,2,3-Propanetriol
TraceCERT™ Analytical Standard
1.252 g/mL (at 25°C)
1 mL
HOCH2CH(OH)CH2OH · aq
500 µg/mL in pyrid ine
92.09 (anhydrous basis)
Ampule
182°C (20 mmHg)
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
ASTM is a registered trademark of American Society for Testing and Materials