Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASTM Colour Reference Standard ASTM D 6045, D1500 (ASTM colour 0.5), MilliporeSigma™ Supelco™

Supplier:  MilliporeSigma Supelco 134290500ML

Catalog No. 11-100-2512

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

Featured Industries: Cleaning Products, Cosmetics, Flavors and Fragrances, Food and Beverages and Personal Care
Specifications

Specifications

8042-47-5
500 mL
Color Reference Standard
Certified Reference Material
>93°C
Safety and Handling

Safety and Handling

ShelfLife : Limited shelf life, expiry date on the label

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit