Learn More
CYDEX PHARMACEUTICALS A LIGAND COMPANY Aseptic-Filled Liquid Captisol | 250ml
Supplier:  CYDEX PHARMACEUTICALS A LIGAND COMPANY LC0C7250
Encompass
Captisol is a patent protected, uniquely modified cyclodextrin, whose chemical structure was rationally designed to enable the creation of new products by significantly improving solubility, stability, bioavailability and dosing of active pharmaceutical ingredients (APIs).
Catalog No. NC1796702