Learn More
Thermo Scientific AS14 PRE-COLUMN
Supplier: Thermo Scientific 046124
IonPac AS14 Analytical Column,Dimensions:4 x 250 mm,Maximum Operating Pressure: 27.6 MPa (4000 psi),Mobile Phase Compatibility: pH 2-11; 0-100% HPLC solvents, Bead Diameter ( micrometer): 9.0 micrometer, Pore Size: 100 angstrom, Crosslinkin