Learn More
Thermo Scientific AS14 A SEPERATOR COLUMN
Supplier: Thermo Scientific 056901
IonPac AS14A-5 micrometer Analytical Column, Dimensions: 3 x 150 mm,Maximum Operating Pressure: 4000 psi, Mobile Phase Compatibility: pH 0 -14;0 -100%HPLC solvents, Bead Diameter: 5 micrometer,Pore Size:100 angstrom, Crosslinking (%DVB): 55