Learn More
Thermo Scientific AS11 ANION COLUMN
Supplier: Thermo Scientific 044076
IonPac AS11 Analytical Column, Size:4 mmx 250 mm, Provides fast analysis of inorganic anions and organic acid anions, including polyphosphates, using hydroxide gradient elution
This item is not returnable.
View return policy