Learn More
Thermo Scientific™ SoftFit-L™ Pipette Tip Reload™ System
Pipette samples with the accuracy and precision you expect from your pipette, every time with Thermo Scientific™ SoftFit-L™ pipette tips. For use with Rainin™ LTS pipettes.
$147.65 - $235.00
Specifications
Filtered | Non-Filtered |
---|---|
Color | Natural |
Clean Claims | DNase, RNase, DNA, ATP, and Endotoxin Free |
Format | Refill |
Tip Style | Universal |
Catalog Number | Mfr. No. | Description | Volume (Metric) | Color Code | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Volume (Metric) | Color Code | Price | Quantity & Availability | |||
21-402-558
|
Thermo Scientific™
3722RI |
20L, Non-Filtered, Non-sterile, Refill | 20 μL | Red |
Pack of 960 for $193.00
Case of 5 PK for $876.00
|
|
|||
21-402-579
|
Thermo Scientific™
3732RI |
300L, Non-Filtered, Non-sterile, Refill | 300 μL | Orange |
Pack of 960 for $193.00
Case of 5 PK for $876.00
|
|
|||
21-402-600
|
Thermo Scientific™
3742RI |
1200L, Non-Filtered, Non-sterile, Refill | 1200 μL | Yellow |
Pack of 768 for $197.00
Case of 4 PK for $713.00
|
|
|||
21-402-569
|
Thermo Scientific™
3752RI |
200L, Non-Filtered, Non-sterile, Refill | 200 μL | Green |
Pack of 960 for $193.00
Case of 5 PK for $876.00
|
|
|||
21-402-589
|
Thermo Scientific™
3782RI |
1000L, Non-Filtered, Non-sterile, Refill | 1000 μL | Blue |
Pack of 768 for $194.00
Case of 4 PK for $702.00
|
|
|||
21-402-556
|
Thermo Scientific™
3721RI |
20L, Non-Filtered, Sterile, Refill | 20 μL | Red |
Pack of 960
for $174.65
Save 26% Reg :
Case of 5 PK
for $790.65
Save 26% Reg : |
|
|||
21-402-577
|
Thermo Scientific™
3731RI |
300L, Non-Filtered, Sterile, Refill | 300 μL | Orange |
Pack of 960 for $233.00
Case of 5 PK for $1,056.00
|
|
|||
21-402-598
|
Thermo Scientific™
3741RI |
1200L, Non-Filtered, Sterile, Refill | 1200 μL | Yellow |
Pack of 768 for $227.00
Case of 4 PK for $821.00
|
|
|||
21-402-567
|
Thermo Scientific™
3751RI |
200L, Non-Filtered, Sterile, Refill | 200 μL | Green |
Pack of 960 for $235.00
Case of 5 PK for $1,064.00
|
|
|||
21-402-587
|
Thermo Scientific™
3781RI |
1000L, Non-Filtered, Sterile, Refill | 1000 μL | Blue |
Pack of 768
for $147.65
Save 32% Reg :
Case of 4 PK
for $532.65
Save 32% Reg : |
|
|||
Description
Pipette samples with the accuracy and precision you expect from your pipette, every time with Thermo Scientific™ SoftFit-L™ pipette tips. For use with Rainin™ LTS pipettes.
The SoftFit-L Reload System is a complete product offering of pipet tips compatible with LTS™ pipetters made by Rainin Instrument, LLC. Designed in response to customer demands, this unique system provides preloaded pipet tips in color-coded inserts by volume size with the option of an easy-to-use “push down” reload pack. Choose sterile or nonsterile tips, in five volume sizes, with or without our proprietary low-retention technology.
- Reload towers are an environmentally-friendly packaging option that reduces plastic waste and necessary storage space
- Compatible with SoftFit-L empty racks
- Color-coded inserts allows quick identification of tip volume
Specifications
Non-Filtered | |
DNase, RNase, DNA, ATP, and Endotoxin Free | |
Universal | |
ART SoftFit L |
Natural | |
Refill | |
Rainin LTS LiteTouch |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.