Learn More
Sartorius arium™ mini UV lamp (185 | 254nm)
For use with arium mini and arium mini plus Ultrapure Water Systems
Supplier: Sartorius H2OCEL1
Description
- Ultrapure water, free of TOC
- Horizontal installation, optimized temperature gradient
- Effective breakdown of organic compounds
- Prevents the growth of microorganisms
- Easy replacement
Specifications
UV Lamp | |
arium mini and arium mini plus Ultrapure Water Systems | |
185/254 nm |